Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PHKG2
- Entrez Gene ID
-
5261
- Gene Name
- phosphorylase kinase catalytic subunit gamma 2
- Gene Aliases
-
GSD9C
- Chromosome Location
-
Chr.16: 30748299 - 30761176 on Build GRCh38
- Gene Symbol
- CCDC189
- Entrez Gene ID
-
90835
- Gene Name
- coiled-coil domain containing 189
- Gene Aliases
-
C16orf93
- Chromosome Location
-
Chr.16: 30757226 - 30762277 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AL122109.2 |
- |
5 - 5 |
|
74 |
View Full Product Details