The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003028.2 | NP_003019.2 | 6 - 6 | 4035 | 71 |
GenBank mRNA | AK055947.1 | - | 1 - 1 | 50 | 71 |