The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177424.2 | NP_803173.1 | 1 - 2 | 248 | 70 |
GenBank mRNA | AF123769.1 | - | 1 - 2 | 195 | 70 |
AK297585.1 | - | 1 - 2 | 176 | 70 | |
AK299982.1 | - | 1 - 2 | 80 | 70 | |
AL035306.1 | - | 1 - 2 | 191 | 70 | |
BC046999.1 | - | 1 - 2 | 176 | 70 | |
CR407620.1 | - | 1 - 2 | 102 | 70 |