The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031800.2 | NP_001026970.1 | 1 - 2 | 338 | 60 |
NM_152902.4 | NP_690866.1 | 1 - 2 | 338 | 60 | |
GenBank mRNA | AB097034.1 | - | 1 - 2 | 221 | 60 |
AK313520.1 | - | 1 - 2 | 338 | 60 | |
AL049670.1 | - | 1 - 2 | 250 | 60 | |
BC009506.2 | - | 1 - 2 | 128 | 60 |