Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MXRA5
Entrez Gene ID
25878
Gene Name
matrix remodeling associated 5
Gene Aliases
-
Chromosome Location
Chr.X: 3308565 - 3347098 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015419.3 NP_056234.2 7 - 7 7541 81
XM_005274485.2 XP_005274542.1 7 - 7 7996 81
GenBank mRNA AF245505.1 - 6 - 6 7384 81
AL049946.1 - 1 - 1 654 81

View Full Product Details