The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256090.1 | NP_001243019.1 | 15 - 15 | 3469 | 133 |
NM_001256091.1 | NP_001243020.1 | 14 - 14 | 3394 | 133 | |
NM_001256092.1 | NP_001243021.1 | 13 - 13 | 3337 | 133 | |
NM_001256093.1 | NP_001243022.1 | 13 - 13 | 3369 | 133 | |
NM_001880.3 | NP_001871.2 | 14 - 14 | 3434 | 133 | |
NR_045768.1 | 17 - 17 | 3631 | 133 | ||
NR_045769.1 | 17 - 17 | 3663 | 133 | ||
NR_045770.1 | 16 - 16 | 3516 | 133 | ||
NR_045771.1 | 15 - 15 | 3508 | 133 | ||
NR_045772.1 | 15 - 15 | 3446 | 133 | ||
NR_045773.1 | 14 - 14 | 3411 | 133 | ||
NR_045774.1 | 13 - 13 | 3319 | 133 | ||
GenBank mRNA | AF283776.2 | - | 1 - 1 | 1055 | 133 |
AK128731.1 | - | 15 - 15 | 2775 | 133 | |
AL050192.1 | - | 1 - 1 | 1047 | 133 | |
BC107698.1 | - | 13 - 13 | 3304 | 133 | |
BX648469.1 | - | 13 - 13 | 3222 | 133 |