Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NPHP4
Entrez Gene ID
261734
Gene Name
nephronophthisis 4
Gene Aliases
POC10, SLSN4
Chromosome Location
Chr.1: 5862808 - 5992473 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291593.1 NP_001278522.1 13 - 14 2102 67
NM_001291594.1 NP_001278523.1 12 - 13 1925 67
NM_015102.4 NP_055917.1 16 - 17 2412 67
NR_111987.1 16 - 17 2409 67
XM_006710563.3 XP_006710626.1 16 - 17 2258 67
XM_011541213.1 XP_011539515.1 15 - 16 2179 67
XM_011541214.1 XP_011539516.1 15 - 16 2182 67
XM_011541215.1 XP_011539517.1 14 - 15 2071 67
XM_011541216.2 XP_011539518.1 17 - 18 2886 67
XM_011541217.2 XP_011539519.1 16 - 17 2750 67
XM_011541218.2 XP_011539520.1 17 - 18 2419 67
XM_017000996.1 XP_016856485.1 15 - 16 2179 67
XM_017000997.1 XP_016856486.1 16 - 17 2681 67
XM_017000998.1 XP_016856487.1 15 - 16 2182 67
XM_017000999.1 XP_016856488.1 16 - 17 2177 67
XM_017001000.1 XP_016856489.1 15 - 16 2010 67
XM_017001001.1 XP_016856490.1 10 - 11 1449 67
XM_017001002.1 XP_016856491.1 15 - 16 2182 67
XM_017001003.1 XP_016856492.1 8 - 9 1240 67
GenBank mRNA AB014573.1 - 11 - 12 1512 67
AF537130.1 - 16 - 17 2410 67
AK124074.1 - 12 - 13 1878 67
AK314326.1 - 13 - 14 2076 67
AL117405.1 - 2 - 3 224 67
AY118228.1 - 16 - 17 2347 67
BC040520.1 - 16 - 17 2410 67

View Full Product Details