The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282447.2 | NP_001269376.1 | 19 - 20 | 2928 | 148 |
NM_025220.4 | NP_079496.1 | 19 - 20 | 2928 | 148 | |
NM_153202.3 | NP_694882.1 | 18 - 19 | 2850 | 148 | |
XM_005260843.1 | XP_005260900.1 | 19 - 20 | 2385 | 148 | |
XM_006723639.1 | XP_006723702.1 | 19 - 20 | 2385 | 148 | |
XM_006723640.1 | XP_006723703.1 | 19 - 20 | 2376 | 148 | |
XM_011529366.1 | XP_011527668.1 | 19 - 20 | 2284 | 148 | |
XM_011529367.1 | XP_011527669.1 | 19 - 20 | 2245 | 148 | |
XM_011529368.1 | XP_011527670.1 | 18 - 19 | 2307 | 148 | |
XM_011529373.1 | XP_011527675.1 | 18 - 19 | 2359 | 148 | |
GenBank mRNA | AB055891.1 | - | 19 - 20 | 2329 | 148 |
AF466287.1 | - | 19 - 20 | 2247 | 148 | |
AK092627.1 | - | 5 - 6 | 1133 | 148 | |
AK123015.1 | - | 15 - 16 | 2952 | 148 | |
AK126487.1 | - | 13 - 14 | 3929 | 148 | |
AK126498.1 | - | 7 - 8 | 3685 | 148 | |
AK226148.1 | - | 9 - 10 | 3992 | 148 | |
AL117415.1 | - | 8 - 9 | 1803 | 148 | |
AY358314.1 | - | 19 - 20 | 2355 | 148 | |
BC125112.1 | - | 7 - 8 | 1393 | 148 | |
BC125113.1 | - | 17 - 18 | 1929 | 148 |