The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145775.2 | NP_001139247.1 | 12 - 12 | 1864 | 70 |
NM_001145776.1 | NP_001139248.1 | 11 - 11 | 1751 | 70 | |
NM_004117.3 | NP_004108.1 | 11 - 11 | 1706 | 70 | |
GenBank mRNA | AF194172.1 | - | 11 - 11 | 1701 | 70 |
AK222807.1 | - | 11 - 11 | 1679 | 70 | |
AK302704.1 | - | 7 - 7 | 1152 | 70 | |
AK303356.1 | - | 11 - 11 | 1568 | 70 | |
AL122066.1 | - | 1 - 1 | 55 | 70 | |
BC042605.1 | - | 12 - 12 | 1864 | 70 | |
BC111050.1 | - | 11 - 11 | 1667 | 70 | |
BX647390.1 | - | 11 - 11 | 1679 | 70 | |
U42031.1 | - | 10 - 10 | 1525 | 70 |