Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C12orf29
Entrez Gene ID
91298
Gene Name
chromosome 12 open reading frame 29
Gene Aliases
-
Chromosome Location
Chr.12: 88035491 - 88050160 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AL137488.1 - 1 - 2 287 84

View Full Product Details