The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172364.4 | NP_758952.4 | 24 - 25 | 2577 | 76 |
XM_011521041.2 | XP_011519343.1 | 23 - 24 | 6416 | 76 | |
GenBank mRNA | AF516695.1 | - | 24 - 25 | 2709 | 76 |
AL137658.1 | - | 2 - 3 | 187 | 76 | |
BC150186.1 | - | 24 - 25 | 2562 | 76 | |
BX537436.1 | - | 24 - 25 | 2540 | 76 | |
BX537437.1 | - | 24 - 25 | 2578 | 76 |