Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IKZF2
Entrez Gene ID
22807
Gene Name
IKAROS family zinc finger 2
Gene Aliases
ANF1A2, HELIOS, ZNF1A2, ZNFN1A2
Chromosome Location
Chr.2: 212999684 - 213152753 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079526.1 NP_001072994.1 9 - 9 7480 148
NM_016260.2 NP_057344.2 8 - 8 7411 148
XM_005246384.4 XP_005246441.1 8 - 8 7521 148
XM_005246385.3 XP_005246442.1 8 - 8 7551 148
XM_005246386.4 XP_005246443.1 8 - 8 7402 148
XM_011510802.2 XP_011509104.1 9 - 9 7593 148
XM_011510803.2 XP_011509105.1 9 - 9 7442 148
XM_011510804.2 XP_011509106.1 9 - 9 7365 148
XM_011510805.2 XP_011509107.1 9 - 9 7454 148
XM_011510807.2 XP_011509109.1 9 - 9 7629 148
XM_011510808.2 XP_011509110.1 9 - 9 7479 148
XM_011510809.2 XP_011509111.1 9 - 9 7491 148
XM_011510810.2 XP_011509112.1 9 - 9 7405 148
XM_011510811.2 XP_011509113.1 8 - 8 7526 148
XM_011510812.2 XP_011509114.1 8 - 8 7387 148
XM_011510814.2 XP_011509116.1 8 - 8 7885 148
XM_011510815.2 XP_011509117.1 8 - 8 7333 148
XM_011510816.2 XP_011509118.1 6 - 6 7334 148
XM_011510817.2 XP_011509119.1 7 - 7 7269 148
XM_011510818.2 XP_011509120.1 6 - 6 12988 148
XM_011510819.2 XP_011509121.1 7 - 7 7258 148
XM_017003589.1 XP_016859078.1 8 - 8 7473 148
XM_017003590.1 XP_016859079.1 8 - 8 7443 148
XM_017003591.1 XP_016859080.1 6 - 6 7876 148
XM_017003592.1 XP_016859081.1 7 - 7 7190 148

View Full Product Details