The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001251825.1 | NP_001238754.1 | 6 - 6 | 5441 | 104 |
NM_003109.1 | NP_003100.1 | 6 - 6 | 5522 | 104 | |
NM_138473.2 | NP_612482.2 | 6 - 6 | 5585 | 104 | |
XM_011538696.2 | XP_011536998.1 | 5 - 5 | 5500 | 104 | |
GenBank mRNA | AL442093.1 | - | 1 - 1 | 451 | 104 |