Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HELQ
Entrez Gene ID
113510
Gene Name
helicase, POLQ-like
Gene Aliases
HEL308
Chromosome Location
Chr.4: 83407343 - 83455883 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AL512702.1 - 1 - 2 32 88

View Full Product Details