The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184723.1 | NP_001171652.1 | 3 - 4 | 499 | 70 |
NM_138786.3 | NP_620141.1 | 4 - 5 | 675 | 70 | |
GenBank mRNA | AK313400.1 | - | 4 - 5 | 449 | 70 |
AL833134.1 | - | 4 - 5 | 597 | 70 | |
BC014339.1 | - | 4 - 5 | 551 | 70 | |
CA454331.1 | - | 3 - 4 | 461 | 70 |