The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134793.1 | NP_001128265.1 | 1 - 2 | 110 | |
XM_017017320.1 | XP_016872809.1 | 1 - 2 | 426 | 111 | |
XM_017017321.1 | XP_016872810.1 | 1 - 2 | 426 | 108 | |
GenBank mRNA | AK127394.1 | - | 1 - 2 | 113 | |
BC015047.1 | - | 1 - 2 | 48 | 111 |