The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281768.1 | NP_001268697.1 | 5 - 6 | 727 | 56 |
NM_021116.2 | NP_066939.1 | 4 - 5 | 1041 | 56 | |
XM_005249584.3 | XP_005249641.1 | 4 - 5 | 1039 | 56 | |
XM_005249585.2 | XP_005249642.1 | 4 - 5 | 1039 | 56 | |
GenBank mRNA | AB209496.1 | - | 4 - 5 | 986 | 56 |
L05500.1 | - | 2 - 3 | 185 | 56 |