The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001293725.1 | NP_001280654.1 | 1 - 2 | 397 | 76 |
NM_005640.2 | NP_005631.1 | 1 - 2 | 397 | 76 | |
NR_121653.1 | 1 - 2 | 397 | 76 | ||
XM_005258339.3 | XP_005258396.1 | 1 - 2 | 789 | 76 | |
XM_011526153.2 | XP_011524455.1 | 1 - 2 | 789 | 76 | |
XM_017025932.1 | XP_016881421.1 | 1 - 2 | 789 | 76 | |
GenBank mRNA | AB234096.1 | - | 1 - 2 | 794 | 76 |
AY888048.1 | - | 1 - 2 | 397 | 76 | |
AY888049.1 | - | 1 - 2 | 397 | 76 | |
Y09321.1 | - | 1 - 2 | 148 | 76 |