Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 22077393-22077902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHEX
Entrez Gene ID
5251
Gene Name
phosphate regulating endopeptidase homolog, X-linked
Gene Aliases
HPDR, HPDR1, HYP, HYP1, LXHR, PEX, XLH
UniGene
Hs.495834
Gene Chromosome Location
Chr.X: 22032327-22251310 on Build GRCh38
Transcripts
NM_000444.5 NM_001282754.1 XM_017029579.1

SNP Detail

SNP ID Chromosome Location
rs750293039 Chr.X: 22077734-22077734 on Build GRCh38
rs749081778 Chr.X: 22077479-22077479 on Build GRCh38
rs763608782 Chr.X: 22077710-22077710 on Build GRCh38
rs759053020 Chr.X: 22077780-22077780 on Build GRCh38
rs748086510 Chr.X: 22077459-22077459 on Build GRCh38
rs756115044 Chr.X: 22077460-22077460 on Build GRCh38
rs140231382 Chr.X: 22077528-22077528 on Build GRCh38
rs946233329 Chr.X: 22077746-22077746 on Build GRCh38
rs778710788 Chr.X: 22077580-22077580 on Build GRCh38
rs1057517790 Chr.X: 22077542-22077542 on Build GRCh38
rs755595286 Chr.X: 22077547-22077547 on Build GRCh38
rs904807446 Chr.X: 22077800-22077800 on Build GRCh38
rs765782770 Chr.X: 22077754-22077754 on Build GRCh38
rs761971488 Chr.X: 22077519-22077519 on Build GRCh38
rs899511782 Chr.X: 22077865-22077865 on Build GRCh38
rs781542537 Chr.X: 22077618-22077618 on Build GRCh38
rs773617103 Chr.X: 22077657-22077657 on Build GRCh38
rs997951924 Chr.X: 22077467-22077467 on Build GRCh38
rs763461759 Chr.X: 22077659-22077659 on Build GRCh38
rs767157065 Chr.X: 22077543-22077543 on Build GRCh38
rs757149988 Chr.X: 22077717-22077717 on Build GRCh38
rs1057518348 Chr.X: 22077704-22077704 on Build GRCh38
rs886041680 Chr.X: 22077547-22077568 on Build GRCh38
rs187824835 Chr.X: 22077544-22077544 on Build GRCh38
rs970161374 Chr.X: 22077469-22077469 on Build GRCh38
rs138497409 Chr.X: 22077521-22077521 on Build GRCh38
rs886039581 Chr.X: 22077577-22077577 on Build GRCh38
rs867854522 Chr.X: 22077782-22077782 on Build GRCh38
rs776784616 Chr.X: 22077516-22077516 on Build GRCh38
rs145881500 Chr.X: 22077483-22077483 on Build GRCh38
rs777607846 Chr.X: 22077557-22077557 on Build GRCh38
rs137961493 Chr.X: 22077576-22077576 on Build GRCh38
rs1057517789 Chr.X: 22077539-22077539 on Build GRCh38
rs751230094 Chr.X: 22077535-22077535 on Build GRCh38
rs899528648 Chr.X: 22077852-22077852 on Build GRCh38
rs754508886 Chr.X: 22077441-22077441 on Build GRCh38
rs374359619 Chr.X: 22077871-22077871 on Build GRCh38
rs765714025 Chr.X: 22077520-22077520 on Build GRCh38
rs777749067 Chr.X: 22077471-22077471 on Build GRCh38
rs748279755 Chr.X: 22077623-22077623 on Build GRCh38
rs753767214 Chr.X: 22077564-22077564 on Build GRCh38
rs372618065 Chr.X: 22077672-22077672 on Build GRCh38
rs763587919 Chr.X: 22077853-22077853 on Build GRCh38
rs771208171 Chr.X: 22077660-22077660 on Build GRCh38
rs886042025 Chr.X: 22077487-22077487 on Build GRCh38
rs886041694 Chr.X: 22077656-22077656 on Build GRCh38
rs769066725 Chr.X: 22077617-22077617 on Build GRCh38
rs778989347 Chr.X: 22077718-22077718 on Build GRCh38
rs747237564 Chr.X: 22077606-22077606 on Build GRCh38
rs780626060 Chr.X: 22077454-22077454 on Build GRCh38
rs770726332 Chr.X: 22077480-22077480 on Build GRCh38
rs370610267 Chr.X: 22077670-22077670 on Build GRCh38
rs36117823 Chr.X: 22077664-22077665 on Build GRCh38
rs775603606 Chr.X: 22077482-22077482 on Build GRCh38
rs774907553 Chr.X: 22077692-22077692 on Build GRCh38
rs774768015 Chr.X: 22077665-22077665 on Build GRCh38
rs766719010 Chr.X: 22077530-22077530 on Build GRCh38
rs150297950 Chr.X: 22077536-22077536 on Build GRCh38
rs758236802 Chr.X: 22077743-22077743 on Build GRCh38
rs768868675 Chr.X: 22077489-22077489 on Build GRCh38
rs769878509 Chr.X: 22077652-22077652 on Build GRCh38
rs376914541 Chr.X: 22077716-22077716 on Build GRCh38
rs771512591 Chr.X: 22077813-22077813 on Build GRCh38

View Full Product Details