Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 107639271-107639802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRPS1
Entrez Gene ID
5631
Gene Name
phosphoribosyl pyrophosphate synthetase 1
Gene Aliases
ARTS, CMTX5, DFN2, DFNX1, PPRibP, PRS-I, PRSI
UniGene
Hs.56
Gene Chromosome Location
Chr.X: 107628424-107651026 on Build GRCh38
Transcripts
NM_001204402.1 NM_002764.3

SNP Detail

SNP ID Chromosome Location
rs760049067 Chr.X: 107639654-107639654 on Build GRCh38
rs755850703 Chr.X: 107639433-107639433 on Build GRCh38
rs866112655 Chr.X: 107639733-107639733 on Build GRCh38
rs780551723 Chr.X: 107639415-107639415 on Build GRCh38
rs372166721 Chr.X: 107639679-107639679 on Build GRCh38
rs1027862985 Chr.X: 107639418-107639418 on Build GRCh38
rs1015038412 Chr.X: 107639563-107639563 on Build GRCh38
rs750054054 Chr.X: 107639421-107639421 on Build GRCh38
rs767243595 Chr.X: 107639420-107639420 on Build GRCh38
rs998598761 Chr.X: 107639616-107639616 on Build GRCh38
rs567661963 Chr.X: 107639749-107639749 on Build GRCh38
rs11797233 Chr.X: 107639573-107639573 on Build GRCh38
rs966136033 Chr.X: 107639666-107639666 on Build GRCh38
rs902859863 Chr.X: 107639599-107639599 on Build GRCh38
rs752194539 Chr.X: 107639460-107639460 on Build GRCh38
rs180177151 Chr.X: 107639365-107639365 on Build GRCh38
rs990453848 Chr.X: 107639736-107639736 on Build GRCh38
rs147896410 Chr.X: 107639349-107639349 on Build GRCh38
rs80338731 Chr.X: 107639301-107639301 on Build GRCh38
rs1023149138 Chr.X: 107639747-107639747 on Build GRCh38
rs758657216 Chr.X: 107639479-107639480 on Build GRCh38
rs773719662 Chr.X: 107639356-107639356 on Build GRCh38
rs876661263 Chr.X: 107639419-107639419 on Build GRCh38
rs766786656 Chr.X: 107639442-107639442 on Build GRCh38
rs755093762 Chr.X: 107639478-107639479 on Build GRCh38
rs371538576 Chr.X: 107639355-107639355 on Build GRCh38
rs777885394 Chr.X: 107639495-107639496 on Build GRCh38
rs137852542 Chr.X: 107639326-107639326 on Build GRCh38
rs1012174319 Chr.X: 107639361-107639361 on Build GRCh38
rs932184130 Chr.X: 107639724-107639724 on Build GRCh38
rs1033217069 Chr.X: 107639619-107639619 on Build GRCh38
rs983928739 Chr.X: 107639424-107639424 on Build GRCh38
rs921933929 Chr.X: 107639707-107639707 on Build GRCh38
rs1029293242 Chr.X: 107639554-107639554 on Build GRCh38
rs180177152 Chr.X: 107639431-107639431 on Build GRCh38

View Full Product Details