The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015175.2 | NP_055990.1 | 45 - 46 | 7317 | 106 |
XM_006713072.3 | XP_006713135.1 | 44 - 45 | 7233 | 106 | |
XM_017006010.1 | XP_016861499.1 | 45 - 46 | 7314 | 106 | |
XM_017006011.1 | XP_016861500.1 | 45 - 46 | 7170 | 106 | |
XM_017006012.1 | XP_016861501.1 | 44 - 45 | 7233 | 106 | |
XM_017006014.1 | XP_016861503.1 | 44 - 45 | 7089 | 106 | |
XM_017006015.1 | XP_016861504.1 | 43 - 44 | 6945 | 106 | |
XM_017006016.1 | XP_016861505.1 | 40 - 41 | 7891 | 106 | |
XM_017006017.1 | XP_016861506.1 | 22 - 23 | 3733 | 106 | |
GenBank mRNA | AB011112.3 | - | 31 - 32 | 5001 | 106 |
AK131104.1 | - | 45 - 46 | 7157 | 106 | |
AK293387.1 | - | 6 - 7 | 688 | 106 | |
AK301269.1 | - | 10 - 11 | 1380 | 106 | |
BC029944.1 | - | 7 - 8 | 738 | 106 | |
BC060874.1 | - | 13 - 14 | 1795 | 106 |