Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GPATCH8
- Entrez Gene ID
-
23131
- Gene Name
- G-patch domain containing 8
- Gene Aliases
-
GPATC8, KIAA0553
- Chromosome Location
-
Chr.17: 44395277 - 44503602 on Build GRCh38
View Full Product Details