Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EXPH5
Entrez Gene ID
23086
Gene Name
exophilin 5
Gene Aliases
SLAC2-B, SLAC2B
Chromosome Location
Chr.11: 108505431 - 108607514 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144763.1 NP_001138235.1 5 - 6 696 65
NM_001144764.1 NP_001138236.1 3 - 4 431 65
NM_001144765.1 NP_001138237.1 2 - 3 382 65
NM_001308019.1 NP_001294948.1 6 - 7 613 65
NM_015065.2 NP_055880.2 5 - 6 740 65
XM_011542696.2 XP_011540998.1 6 - 7 634 65
XM_011542698.2 XP_011541000.1 5 - 6 1080 65
XM_011542700.2 XP_011541002.1 5 - 6 1980 65
XM_017017397.1 XP_016872886.1 6 - 7 629 65
XM_017017398.1 XP_016872887.1 6 - 7 626 65
XM_017017399.1 XP_016872888.1 5 - 6 532 65
XM_017017400.1 XP_016872889.1 6 - 7 2048 65
XM_017017401.1 XP_016872890.1 5 - 6 1977 65
XM_017017402.1 XP_016872891.1 6 - 7 2088 65
XM_017017403.1 XP_016872892.1 3 - 4 1234 65
XM_017017404.1 XP_016872893.1 4 - 5 1931 65
GenBank mRNA AB014524.1 - 6 - 7 613 65
AK303993.1 - 3 - 4 411 65
AK304212.1 - 2 - 3 383 65
AY099469.1 - 5 - 6 726 65
BC113119.1 - 5 - 6 711 65
BC117701.1 - 5 - 6 711 65
CR627226.1 - 5 - 6 702 65
DC415184.1 - 3 - 4 411 65
DC416995.1 - 2 - 3 383 65

View Full Product Details