The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172735.2 | NP_001166206.1 | 9 - 10 | 1268 | 84 |
NM_015568.3 | NP_056383.1 | 10 - 11 | 1394 | 84 | |
XM_011528768.2 | XP_011527070.1 | 9 - 10 | 1957 | 84 | |
XM_011528769.2 | XP_011527071.1 | 8 - 9 | 975 | 84 | |
XM_017027785.1 | XP_016883274.1 | 9 - 10 | 1289 | 84 | |
GenBank mRNA | AB020630.2 | - | 10 - 11 | 1382 | 84 |
AB177855.1 | - | 8 - 9 | 895 | 84 | |
AF362910.1 | - | 9 - 10 | 1245 | 84 | |
BC131801.1 | - | 10 - 11 | 1351 | 84 | |
BC152467.1 | - | 10 - 11 | 1382 | 84 |