Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STXBP5L
Entrez Gene ID
9515
Gene Name
syntaxin binding protein 5 like
Gene Aliases
LLGL4
Chromosome Location
Chr.3: 120908203 - 121424761 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308330.1 NP_001295259.1 4 - 5 510 122
NM_014980.2 NP_055795.1 4 - 5 510 122
XM_006713825.3 XP_006713888.1 4 - 5 680 122
XM_011513328.2 XP_011511630.1 3 - 4 357 122
XM_011513331.2 XP_011511633.1 4 - 5 507 122
XM_011513333.2 XP_011511635.1 4 - 5 507 122
XM_017007530.1 XP_016863019.1 4 - 5 441 122
XM_017007531.1 XP_016863020.1 4 - 5 495 122
XM_017007532.1 XP_016863021.1 4 - 5 444 122
XM_017007534.1 XP_016863023.1 4 - 5 507 122
XM_017007535.1 XP_016863024.1 4 - 5 698 122
GenBank mRNA AB023223.1 - 4 - 5 477 122
AK296544.1 - 4 - 5 508 122

View Full Product Details