The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052964.2 | NP_443196.2 | 11 - 12 | 743 | 80 |
XM_011513775.2 | XP_011512077.1 | 11 - 12 | 974 | 80 | |
XM_017007684.1 | XP_016863173.1 | 11 - 12 | 974 | 80 | |
XM_017007685.1 | XP_016863174.1 | 11 - 12 | 974 | 80 | |
GenBank mRNA | AB032369.1 | - | 9 - 10 | 552 | 80 |
AB110420.1 | - | 11 - 12 | 846 | 80 | |
AK093001.1 | - | 11 - 12 | 745 | 80 | |
BC137553.1 | - | 11 - 12 | 672 | 80 | |
BC137556.1 | - | 11 - 12 | 672 | 80 |