The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020746.4 | NP_065797.2 | 2 - 3 | 291 | 71 |
NR_037921.1 | 2 - 3 | 291 | 71 | ||
GenBank mRNA | AB033097.1 | - | 1 - 2 | 197 | 71 |
AB232371.1 | - | 1 - 2 | 191 | 71 | |
AK023799.1 | - | 2 - 3 | 247 | 71 | |
AK123956.1 | - | 2 - 3 | 252 | 71 | |
AK130992.1 | - | 2 - 3 | 212 | 71 | |
AK296897.1 | - | 2 - 3 | 256 | 71 | |
BC044952.1 | - | 2 - 3 | 225 | 71 | |
BM834250.1 | - | 2 - 3 | 255 | 71 | |
DB219666.1 | - | 2 - 3 | 291 | 71 | |
DQ167126.1 | - | 1 - 2 | 119 | 71 | |
DQ174270.1 | - | 1 - 2 | 119 | 71 | |
DQ181928.1 | - | 1 - 2 | 119 | 71 | |
EF467324.1 | - | 1 - 2 | 119 | 71 | |
KC415005.1 | - | 1 - 2 | 119 | 71 |