Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBM27
Entrez Gene ID
54439
Gene Name
RNA binding motif protein 27
Gene Aliases
ARRS1, Psc1, ZC3H18, ZC3H20
Chromosome Location
Chr.5: 146203550 - 146289223 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018989.1 NP_061862.1 8 - 9 1445 63
XM_005268466.1 XP_005268523.1 8 - 9 1498 63
XM_006714787.1 XP_006714850.1 9 - 10 1462 63
XM_011537661.1 XP_011535963.1 8 - 9 1498 63
XM_011537662.1 XP_011535964.1 8 - 9 1495 63
XM_017009587.1 XP_016865076.1 9 - 10 1459 63
GenBank mRNA AB037732.1 - 4 - 5 768 63
BX647384.1 - 8 - 9 1445 63

View Full Product Details