The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020768.3 | NP_065819.1 | 3 - 4 | 1467 | 70 |
XM_005268492.2 | XP_005268549.1 | 2 - 3 | 1858 | 70 | |
XM_005268493.2 | XP_005268550.1 | 2 - 3 | 1254 | 70 | |
XM_006714789.2 | XP_006714852.1 | 2 - 3 | 1918 | 70 | |
XM_011537671.2 | XP_011535973.1 | 4 - 5 | 1753 | 70 | |
GenBank mRNA | AB037738.1 | - | 2 - 3 | 1929 | 70 |
AK292477.1 | - | 3 - 4 | 1508 | 70 | |
BC111962.1 | - | 1 - 2 | 860 | 70 | |
BC113435.1 | - | 1 - 2 | 860 | 70 | |
CR749266.1 | - | 3 - 4 | 1468 | 70 |