The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020872.2 | NP_065923.1 | 7 - 8 | 977 | 59 |
XM_005264757.3 | XP_005264814.1 | 8 - 9 | 1087 | 59 | |
XM_011533768.2 | XP_011532070.1 | 7 - 8 | 4609 | 59 | |
XM_017006507.1 | XP_016861996.1 | 8 - 9 | 1657 | 59 | |
XM_017006508.1 | XP_016861997.1 | 7 - 8 | 882 | 59 | |
GenBank mRNA | AB040929.1 | - | 5 - 6 | 626 | 59 |
BC150608.1 | - | 7 - 8 | 977 | 59 |