The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033553.2 | NP_001028725.1 | 2 - 3 | 195 | 90 |
NM_001243439.1 | NP_001230368.1 | 2 - 3 | 254 | 90 | |
NM_152904.4 | NP_690868.3 | 2 - 3 | 195 | 90 | |
XM_011524075.1 | XP_011522377.1 | 1 - 2 | 147 | 90 | |
XM_017025317.1 | XP_016880806.1 | 1 - 2 | 147 | 90 | |
XM_017025318.1 | XP_016880807.1 | 1 - 2 | 147 | 90 | |
XM_017025319.1 | XP_016880808.1 | 2 - 3 | 289 | 90 | |
XM_017025320.1 | XP_016880809.1 | 2 - 3 | 289 | 90 | |
XM_017025322.1 | XP_016880811.1 | 1 - 2 | 147 | 90 | |
XM_017025323.1 | XP_016880812.1 | 1 - 2 | 147 | 90 | |
XM_017025324.1 | XP_016880813.1 | 1 - 2 | 147 | 90 | |
XM_017025325.1 | XP_016880814.1 | 2 - 3 | 289 | 90 | |
GenBank mRNA | AB041533.1 | - | 2 - 3 | 195 | 90 |
AK295093.1 | - | 2 - 3 | 227 | 90 | |
AK314341.1 | - | 2 - 3 | 270 | 90 | |
AY816328.1 | - | 2 - 3 | 195 | 90 | |
AY816329.1 | - | 2 - 3 | 195 | 90 | |
DA058164.1 | - | 2 - 3 | 290 | 90 |