The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020935.2 | NP_065986.2 | 5 - 6 | 741 | 90 |
XM_005246720.2 | XP_005246777.1 | 5 - 6 | 813 | 90 | |
XM_005246721.3 | XP_005246778.1 | 5 - 6 | 705 | 90 | |
XM_005246722.3 | XP_005246779.1 | 5 - 6 | 745 | 90 | |
XM_005246724.1 | XP_005246781.1 | 5 - 6 | 741 | 90 | |
XM_011511538.2 | XP_011509840.1 | 4 - 5 | 686 | 90 | |
XM_011511541.2 | XP_011509843.1 | 5 - 6 | 741 | 90 | |
XM_017004577.1 | XP_016860066.1 | 5 - 6 | 699 | 90 | |
GenBank mRNA | AB046814.1 | - | 1 - 2 | 186 | 90 |
AK095577.1 | - | 5 - 6 | 699 | 90 | |
AK128583.1 | - | 5 - 6 | 745 | 90 | |
AL832645.2 | - | 5 - 6 | 740 | 90 | |
BC050461.1 | - | 5 - 6 | 710 | 90 | |
BC112901.1 | - | 4 - 5 | 541 | 90 | |
BC133007.1 | - | 5 - 6 | 626 | 90 | |
BC133009.1 | - | 5 - 6 | 671 | 90 | |
BC144249.1 | - | 5 - 6 | 626 | 90 | |
BC144252.1 | - | 5 - 6 | 666 | 90 | |
BX538024.1 | - | 5 - 6 | 706 | 90 |