Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TRPM3
- Entrez Gene ID
-
80036
- Gene Name
- transient receptor potential cation channel subfamily M member 3
- Gene Aliases
-
GON-2, LTRPC3, MLSN2
- Chromosome Location
-
Chr.9: 70529063 - 71446989 on Build GRCh38
View Full Product Details