Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZFYVE28
Entrez Gene ID
57732
Gene Name
zinc finger FYVE-type containing 28
Gene Aliases
LST2, LYST2
Chromosome Location
Chr.4: 2269585 - 2418698 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172656.1 NP_001166127.1 1 - 2 359 82
NM_001172657.1 NP_001166128.1 1 - 2 359 82
NM_020972.2 NP_066023.2 1 - 2 359 82
XM_006713900.3 XP_006713963.1 1 - 2 413 82
XM_006713902.3 XP_006713965.1 1 - 2 414 82
XM_006713904.3 XP_006713967.1 1 - 2 413 82
XM_011513541.2 XP_011511843.1 1 - 2 413 82
XM_017008503.1 XP_016863992.1 1 - 2 414 82
XM_017008504.1 XP_016863993.1 1 - 2 413 82
XM_017008506.1 XP_016863995.1 1 - 2 414 82
XM_017008507.1 XP_016863996.1 1 - 2 413 82
XM_017008508.1 XP_016863997.1 1 - 2 413 82
XM_017008509.1 XP_016863998.1 1 - 2 413 82
XM_017008510.1 XP_016863999.1 1 - 2 413 82
GenBank mRNA AB046863.1 - 1 - 2 358 82
AK294710.1 - 2 - 3 186 82
BC053664.1 - 1 - 2 325 82
BC137309.1 - 2 - 3 102 82
BC137310.1 - 2 - 3 102 82

View Full Product Details