Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SLC35G5
- Entrez Gene ID
-
83650
- Gene Name
- solute carrier family 35 member G5
- Gene Aliases
-
AMAC, AMAC1L2
- Chromosome Location
-
Chr.8: 11330986 - 11332186 on Build GRCh38
- Gene Symbol
- SLC35G3
- Entrez Gene ID
-
146861
- Gene Name
- solute carrier family 35 member G3
- Gene Aliases
-
AMAC1, TMEM21A
- Chromosome Location
-
Chr.17: 35192520 - 35194393 on Build GRCh38
- Gene Symbol
- SLC35G6
- Entrez Gene ID
-
643664
- Gene Name
- solute carrier family 35 member G6
- Gene Aliases
-
AMAC1L3, TMEM21B
- Chromosome Location
-
Chr.17: 7481402 - 7483064 on Build GRCh38
View Full Product Details