The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122842.2 | NP_001116314.1 | 10 - 11 | 2435 | 83 |
NM_001199619.1 | NP_001186548.1 | 11 - 12 | 2451 | 83 | |
NM_001199620.1 | NP_001186549.1 | 12 - 13 | 2569 | 83 | |
NM_001199621.1 | NP_001186550.1 | 9 - 10 | 2214 | 83 | |
NM_181782.4 | NP_861447.3 | 10 - 11 | 2468 | 83 | |
XM_005266822.4 | XP_005266879.1 | 10 - 11 | 2498 | 83 | |
XM_006715340.3 | XP_006715403.1 | 10 - 11 | 2425 | 83 | |
XM_011535455.2 | XP_011533757.1 | 10 - 11 | 2373 | 83 | |
XM_017010269.1 | XP_016865758.1 | 10 - 11 | 2184 | 83 | |
XM_017010270.1 | XP_016865759.1 | 11 - 12 | 2397 | 83 | |
XM_017010271.1 | XP_016865760.1 | 10 - 11 | 2395 | 83 | |
XM_017010272.1 | XP_016865761.1 | 10 - 11 | 2248 | 83 | |
XM_017010273.1 | XP_016865762.1 | 10 - 11 | 2465 | 83 | |
XM_017010274.1 | XP_016865763.1 | 10 - 11 | 2554 | 83 | |
GenBank mRNA | AF493978.1 | - | 11 - 12 | 2451 | 83 |
AJ420542.1 | - | 2 - 3 | 401 | 83 | |
AK127512.1 | - | 10 - 11 | 2245 | 83 | |
AK294558.1 | - | 9 - 10 | 2012 | 83 | |
AL832621.2 | - | 10 - 11 | 2227 | 83 | |
AL832628.2 | - | 10 - 11 | 2192 | 83 | |
AL834442.1 | - | 8 - 9 | 1856 | 83 | |
BC033612.1 | - | 3 - 4 | 1306 | 83 | |
BC137094.1 | - | 12 - 13 | 2485 | 83 | |
BC137095.1 | - | 11 - 12 | 2367 | 83 | |
BX537385.1 | - | 10 - 11 | 2263 | 83 |