The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042601.2 | NP_001036066.1 | 9 - 10 | 1301 | 135 |
NM_001288582.1 | NP_001275511.1 | 9 - 10 | 1301 | 135 | |
NM_133462.3 | NP_597719.1 | 9 - 10 | 1301 | 135 | |
XM_011512439.2 | XP_011510741.1 | 5 - 6 | 543 | 135 | |
XM_017005739.1 | XP_016861228.1 | 8 - 9 | 1132 | 135 | |
XM_017005740.1 | XP_016861229.1 | 9 - 10 | 1288 | 135 | |
GenBank mRNA | AB075860.2 | - | 7 - 8 | 839 | 135 |
AK055984.1 | - | 9 - 10 | 1259 | 135 | |
AL832052.1 | - | 9 - 10 | 1239 | 135 | |
AL833086.2 | - | 9 - 10 | 1238 | 135 | |
AL833103.1 | - | 9 - 10 | 1220 | 135 | |
AY358762.1 | - | 9 - 10 | 1221 | 135 | |
BC027990.1 | - | 9 - 10 | 1234 | 135 | |
BX648669.1 | - | 4 - 5 | 3723 | 135 |