Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SDK1
Entrez Gene ID
221935
Gene Name
sidekick cell adhesion molecule 1
Gene Aliases
-
Chromosome Location
Chr.7: 3301448 - 4269000 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079653.1 NP_001073121.1 18 - 19 2667 57
NM_152744.3 NP_689957.3 44 - 45 6522 57
XM_011515188.1 XP_011513490.1 38 - 39 5363 57
XM_011515189.1 XP_011513491.1 38 - 39 5360 57
XM_017011837.1 XP_016867326.1 38 - 39 5363 57
GenBank mRNA AK055987.1 - 5 - 6 593 57
AK074077.1 - 11 - 12 1462 57
AK074083.1 - 29 - 30 4158 57
AL833354.1 - 18 - 19 2664 57
AY310398.1 - 44 - 45 6522 57

View Full Product Details