The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278549.1 | NP_001265478.1 | 12 - 12 | 3270 | 97 |
NM_002610.4 | NP_002601.1 | 11 - 11 | 3210 | 97 | |
NR_103729.1 | 12 - 12 | 1621 | 97 | ||
XM_006712595.3 | XP_006712658.1 | 9 - 9 | 2535 | 97 | |
XM_011511343.2 | XP_011509645.1 | 12 - 12 | 3032 | 97 | |
XM_011511344.2 | XP_011509646.1 | 12 - 12 | 2917 | 97 | |
XM_011511345.2 | XP_011509647.1 | 11 - 11 | 2972 | 97 | |
XM_011511347.2 | XP_011509649.1 | 12 - 12 | 2917 | 97 | |
GenBank mRNA | AK056110.1 | - | 1 - 1 | 956 | 97 |
BC039158.1 | - | 12 - 12 | 1482 | 97 |