The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032664.3 | NP_116053.3 | 4 - 5 | 1570 | 59 |
XM_011544676.2 | XP_011542978.1 | 5 - 6 | 2042 | 59 | |
XM_011544677.2 | XP_011542979.1 | 4 - 5 | 1568 | 59 | |
XM_011544678.2 | XP_011542980.1 | 5 - 6 | 1575 | 59 | |
XM_011544679.2 | XP_011542981.1 | 5 - 6 | 2038 | 59 | |
XM_011544680.2 | XP_011542982.1 | 5 - 6 | 2036 | 59 | |
XM_011544681.2 | XP_011542983.1 | 5 - 6 | 2037 | 59 | |
GenBank mRNA | AJ431184.1 | - | 3 - 4 | 1382 | 59 |
AJ582015.1 | - | 4 - 5 | 1570 | 59 | |
AK056199.1 | - | 1 - 2 | 56 | 59 | |
AK292993.1 | - | 4 - 5 | 1568 | 59 | |
AK297132.1 | - | 5 - 6 | 1663 | 59 | |
AK303231.1 | - | 4 - 5 | 1503 | 59 | |
BC063462.1 | - | 4 - 5 | 1610 | 59 |