Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MB21D2
Entrez Gene ID
151963
Gene Name
Mab-21 domain containing 2
Gene Aliases
C3orf59
Chromosome Location
Chr.3: 192796815 - 192918161 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178496.3 NP_848591.2 2 - 2 753 78
GenBank mRNA AK056276.1 - 1 - 1 149 78
AK290447.1 - 2 - 2 448 78
BC013123.1 - 2 - 2 431 78
BC036194.1 - 2 - 2 737 78
BC045582.1 - 2 - 2 753 78
BX648517.1 - 2 - 2 452 78

View Full Product Details