Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
XRRA1
Entrez Gene ID
143570
Gene Name
X-ray radiation resistance associated 1
Gene Aliases
-
Chromosome Location
Chr.11: 74840910 - 74949187 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270380.1 NP_001257309.1 1 - 2 258 84
NM_182969.2 NP_892014.1 1 - 2 258 84
XM_005273765.2 XP_005273822.1 1 - 2 258 84
XM_011544755.1 XP_011543057.1 1 - 2 258 84
XM_011544756.1 XP_011543058.1 1 - 2 258 84
XM_011544757.1 XP_011543059.1 1 - 2 258 84
XM_011544758.1 XP_011543060.1 1 - 2 258 84
XM_011544759.1 XP_011543061.1 1 - 2 258 84
XM_011544760.2 XP_011543062.1 1 - 2 258 84
XM_011544761.1 XP_011543063.1 1 - 2 258 84
XM_011544762.1 XP_011543064.1 1 - 2 258 84
XM_011544763.1 XP_011543065.1 1 - 2 258 84
XM_011544764.1 XP_011543066.1 1 - 2 258 84
XM_011544766.2 XP_011543068.1 1 - 2 251 84
XM_011544767.1 XP_011543069.1 1 - 2 251 84
XM_017017221.1 XP_016872710.1 1 - 2 258 84
XM_017017222.1 XP_016872711.1 1 - 2 258 84
XM_017017223.1 XP_016872712.1 1 - 2 258 84
XM_017017224.1 XP_016872713.1 1 - 2 258 84
XM_017017225.1 XP_016872714.1 1 - 2 258 84
XM_017017226.1 XP_016872715.1 1 - 2 251 84
XM_017017227.1 XP_016872716.1 1 - 2 251 84
XM_017017229.1 XP_016872718.1 1 - 2 251 84
XM_017017230.1 XP_016872719.1 1 - 2 251 84
XM_017017231.1 XP_016872720.1 1 - 2 251 84
XM_017017233.1 XP_016872722.1 1 - 2 251 84
GenBank mRNA AK056364.1 - 1 - 2 138 84
BC064584.1 - 1 - 2 187 84
DA104497.1 - 1 - 2 202 84
DB040217.1 - 1 - 2 175 83
DB082524.1 - 1 - 2 259 84

View Full Product Details