The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001305203.1 | NP_001292132.1 | 5 - 5 | 1765 | 93 |
NM_001305206.1 | NP_001292135.1 | 5 - 5 | 1820 | 93 | |
NM_001305207.1 | NP_001292136.1 | 5 - 5 | 1817 | 93 | |
NM_001305208.1 | NP_001292137.1 | 5 - 5 | 1810 | 93 | |
NM_133458.3 | NP_597715.2 | 5 - 5 | 1755 | 93 | |
NR_130976.1 | 5 - 5 | 1863 | 93 | ||
NR_130977.1 | 5 - 5 | 1813 | 93 | ||
NR_130978.1 | 5 - 5 | 1871 | 93 | ||
XM_005255804.2 | XP_005255861.1 | 5 - 5 | 1695 | 93 | |
XM_017022952.1 | XP_016878441.1 | 4 - 4 | 1802 | 93 | |
GenBank mRNA | AB075834.1 | - | 1 - 1 | 959 | 93 |
AK056596.1 | - | 1 - 1 | 548 | 93 | |
AK074332.1 | - | 5 - 5 | 1779 | 93 | |
AK122847.1 | - | 1 - 1 | 2190 | 93 | |
AK304152.1 | - | 5 - 5 | 1657 | 93 | |
AK316251.1 | - | 5 - 5 | 1682 | 93 | |
BC033165.1 | - | 1 - 1 | 452 | 93 | |
BC040486.1 | - | 5 - 5 | 1765 | 93 | |
BC140886.1 | - | 5 - 5 | 1755 | 93 | |
BC140887.1 | - | 5 - 5 | 1755 | 93 |