The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300838.1 | NP_001287767.1 | 5 - 6 | 785 | 69 |
NM_001300839.1 | NP_001287768.1 | 5 - 6 | 531 | 69 | |
NM_001300840.1 | NP_001287769.1 | 5 - 6 | 479 | 69 | |
NM_001300841.1 | NP_001287770.1 | 5 - 6 | 547 | 69 | |
NM_144697.3 | NP_653298.1 | 4 - 5 | 1374 | 69 | |
XM_017000378.1 | XP_016855867.1 | 4 - 5 | 640 | 69 | |
GenBank mRNA | AK056960.1 | - | 4 - 5 | 655 | 69 |
AK315397.1 | - | 4 - 5 | 1059 | 69 | |
BC017397.1 | - | 5 - 6 | 503 | 69 | |
BC027999.1 | - | 5 - 6 | 775 | 69 | |
BC047238.1 | - | 4 - 5 | 1088 | 69 | |
BE784098.1 | - | 5 - 6 | 477 | 69 | |
CN359562.1 | - | 5 - 6 | 445 | 69 |