Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CCNY
- Entrez Gene ID
-
219771
- Gene Name
- cyclin Y
- Gene Aliases
-
C10orf9, CBCP1, CCNX, CFP1
- Chromosome Location
-
Chr.10: 35246981 - 35572670 on Build GRCh38
View Full Product Details