The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207307.2 | NP_997190.1 | 8 - 9 | 1620 | 67 |
XM_011513293.2 | XP_011511595.1 | 8 - 9 | 3694 | 67 | |
XM_011513294.2 | XP_011511596.1 | 8 - 9 | 3692 | 67 | |
XM_011513295.2 | XP_011511597.1 | 8 - 9 | 3650 | 67 | |
XM_011513297.2 | XP_011511599.1 | 7 - 8 | 3542 | 67 | |
XM_011513298.1 | XP_011511600.1 | 5 - 6 | 1125 | 67 | |
XM_011513299.2 | XP_011511601.1 | 5 - 6 | 1319 | 67 | |
GenBank mRNA | AK057281.1 | - | 1 - 2 | 1408 | 67 |
AK096099.1 | - | 5 - 6 | 2716 | 67 | |
AK127968.1 | - | 8 - 9 | 1605 | 67 | |
AL133011.1 | - | 6 - 7 | 5074 | 67 | |
BC066975.1 | - | 8 - 9 | 1620 | 67 |