Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC42
Entrez Gene ID
146849
Gene Name
coiled-coil domain containing 42
Gene Aliases
CCDC42A
Chromosome Location
Chr.17: 8729928 - 8744836 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144681.2 NP_653282.2 4 - 5 724 99
GenBank mRNA AK057296.1 - 4 - 5 724 99

View Full Product Details