Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C15orf32
Entrez Gene ID
145858
Gene Name
chromosome 15 open reading frame 32
Gene Aliases
-
Chromosome Location
Chr.15: 92465762 - 92501117 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153040.2 NP_694585.1 2 - 3 963 82
GenBank mRNA AK057393.1 - 2 - 3 964 82
BC109253.1 - 2 - 3 717 82

View Full Product Details