Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TANGO2
Entrez Gene ID
128989
Gene Name
transport and golgi organization 2 homolog
Gene Aliases
C22orf25, MECRCN
Chromosome Location
Chr.22: 20017000 - 20067164 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283129.2 NP_001270058.1 2 - 3 401 76
NM_001283215.2 NP_001270144.1 2 - 3 401 76
NM_001322141.1 NP_001309070.1 2 - 3 401 76
NM_001322143.1 NP_001309072.1 2 - 3 474 76
NM_001322144.1 NP_001309073.1 2 - 3 401 76
NM_001322145.1 NP_001309074.1 2 - 3 474 76
NM_001322147.1 NP_001309076.1 2 - 3 401 76
NM_001322148.1 NP_001309077.1 2 - 3 401 76
NM_001322149.1 NP_001309078.1 2 - 3 401 76
NM_001322150.1 NP_001309079.1 2 - 3 401 76
NM_001322172.1 NP_001309101.1 2 - 3 526 76
NM_001322173.1 NP_001309102.1 2 - 3 401 76
NR_104275.2 2 - 3 401 76
XM_011529865.1 XP_011528167.1 1 - 2 177 76
XM_017028584.1 XP_016884073.1 2 - 3 381 76
GenBank mRNA AK295210.1 - 1 - 2 287 76
AK301366.1 - 2 - 3 360 76
AK309398.1 - 1 - 2 41 76
AK316056.1 - 2 - 3 381 76
BP193987.1 - 2 - 3 359 76
DC332363.1 - 2 - 3 381 76
DC349456.1 - 2 - 3 357 76

View Full Product Details