Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCST2
Entrez Gene ID
127579
Gene Name
DC-STAMP domain containing 2
Gene Aliases
-
Chromosome Location
Chr.1: 155018513 - 155033781 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144622.2 NP_653223.2 8 - 9 1423 97
XM_011509188.2 XP_011507490.1 6 - 7 886 97
XM_011509189.2 XP_011507491.1 3 - 4 1275 97
GenBank mRNA AK057496.1 - 8 - 9 1423 97
AK097468.1 - 6 - 7 837 97
AK097993.1 - 3 - 4 1095 97
BC111794.1 - 8 - 9 1402 97

View Full Product Details